Assay Details
| Gene Symbol: ERP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006817.3 | NP_006808.1 | 1 - 2 | 258 | 133 | |
| GenBank mRNA | BC101493.1 | - | 1 - 2 | 198 | 8068999 | 133 |
| BC101495.1 | - | 1 - 2 | 198 | 8069001 | 133 | |
| CR541667.1 | - | 1 - 2 | 140 | 133 | ||
| FJ224317.1 | - | 1 - 2 | 151 | 133 | ||
| X94910.1 | - | 1 - 2 | 151 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000874.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
positive regulation of protein phosphorylation
protein folding
intracellular protein transport
protein secretion
positive regulation of gene expression
negative regulation of gene expression
protein unfolding
negative regulation of protein secretion
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein disulfide isomerase activity
protein homodimerization activity
chaperone binding
activation of MAPK activity
positive regulation of protein phosphorylation
protein folding
intracellular protein transport
protein secretion
positive regulation of gene expression
negative regulation of gene expression
protein unfolding
negative regulation of protein secretion
regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
protein disulfide isomerase activity
protein homodimerization activity
chaperone binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009470.hg.1 | ENST00000442902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011528.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011528.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011528.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011528.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011528.hg.1 | TC12000874.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011531.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011531.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011531.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011531.hg.1 | TC12000874.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011529.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011529.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011529.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011529.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011529.hg.1 | TC12000874.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166035.hg.1 | TC1200008900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC2000009470.hg.1 | ENST00000442902.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000546477.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008900.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166027.hg.1 | TC1200008900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166029.hg.1 | TC1200008900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000261735.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000455836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000552052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | ENST00000553161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | NM_001034025.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | NM_006817.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166031.hg.1 | TC1200008900.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 140797 | - | - |

ERP29
Genomic Map