Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000433966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010106.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000431296.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
ENST00000458682.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
NR_026795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000168778.hg.1 |
TC1000010106.hg.1 |
Clariom D TC |