Assay Details
Gene Symbol: PRKY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | Y15801.1 | - | 8 | 6039 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2100006799.hg.1 | ENST00000384075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013120.hg.1 | ENST00000384386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008241.hg.1 | ENST00000517049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000495163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000495163.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000516346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009305.hg.1 | ENST00000383871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008241.hg.1 | ENST00000517049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009305.hg.1 | ENST00000383871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000472666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000495163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000495163.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000516346.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000528056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | ENST00000533551.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0Y00006528.hg.1 | NR_028062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100006799.hg.1 | ENST00000384075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013120.hg.1 | ENST00000384386.1 | Ensembl |
