Assay Details
Gene Symbol: KRT6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173086.4 | NP_775109.2 | 9 | 2175 | 139 | |
GenBank mRNA | BC110639.1 | - | 9 | 2175 | 4754834 | 139 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789483 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789442 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354537 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354534 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354551 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789503 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354548 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16558826 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789508 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16558827 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789510 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354545 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08354539 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16558825 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28789443 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 679474 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 242897 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 352829 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 155809 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 223740 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 409149 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 416473 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 346463 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P366936 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P366936 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 35781 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57058 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P33202357 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P39674397 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P11445785 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P366936 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P366936 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789507 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789502 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789507 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354548 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354551 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789483 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789510 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789508 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08354547 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28789505 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P366936 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354548 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354558 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354552 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354550 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354556 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08354546 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P366936 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 7260 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 6515 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P11445785 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354547 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354529 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354556 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16558824 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354526 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354540 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354533 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354552 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354537 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16558877 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354558 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354554 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354525 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354536 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354545 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354542 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08354551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7963416 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963411 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963418 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963419 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963420 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963412 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963415 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963413 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963414 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7963417 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7963410 | - | - |
Affymetrix Human Genome U219 Array | 11727493_x_at | - | - |
Affymetrix Human Genome U219 Array | 11727492_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11727493_PM_x_at | - | - |
Affymetrix Human Genome U219 Array | 11727492_at | - | - |
Human Genome U133 Plus 2.0 Array | 214580_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 214580_x_at | - | - |
Human Genome U133A 2.0 Array | 214580_x_at | - | - |
Human Genome U133A 2.0 Array | 214580_PM_x_at | - | - |
Human Genome U95 Set | 39015_PM_f_at | - | - |
Human Genome U95 Set | 39016_r_at | - | - |
Human Genome U95 Set | 39016_PM_r_at | - | - |
Human Genome U95 Set | 39015_f_at | - | - |
Human Genome U95 Set | 62722_f_at | - | - |
Human Genome U95 Set | 62722_PM_f_at | - | - |
Human Genome U95 Set | 62723_PM_r_at | - | - |
Human Genome U95 Set | 62723_r_at | - | - |
Human Genome U133 Set | 214580_x_at | - | - |
Human Genome U133 Set | 214580_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047608 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047608 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1754576 | L42611.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1754576 | AK299478.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1754576 | BC110639.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31343353-S | L42611.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31343353-S | AK299478.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31343353-S | BC110639.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_31343353-S | NM_173086.4 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_1845-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0003450438 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0003450438 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 460730 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_1845 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754576 | NM_173086.4 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754576 | L42611.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754576 | AK299478.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754576 | BC110639.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754576 | NM_173086.4 | human_transcriptome |
