Assay Details
| Gene Symbol: FREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207361.5 | NP_997244.4 | 18 - 19 | 8592 | 80 | |
| GenBank mRNA | BX538150.1 | - | 1 - 2 | 59 | 80 | |
| BX538304.1 | - | 14 - 15 | 2556 | 80 | ||
| CR933724.1 | - | 6 - 7 | 1084 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000132.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001539.hg.1 | ENST00000280481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001539.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13001539.hg.1 | TC13000132.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | ENST00000482551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300006919.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147022.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147022.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147022.hg.1 | ENST00000280481.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300147022.hg.1 | NM_207361.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300147022.hg.1 | TC1300006919.hg.1 | Clariom D TC |

FREM2
Genomic Map