Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000438.hg.1 |
TC01000035.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000438.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01000438.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006502.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144770.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144770.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144770.hg.1 |
ENST00000339381.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100144770.hg.1 |
NM_152228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100144770.hg.1 |
TC0100006502.hg.1 |
Clariom D TC |