Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207695.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207695.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207695.hg.1 |
TC0100014377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207699.hg.1 |
TC0100014377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207698.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207698.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207698.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207698.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207698.hg.1 |
TC0100014377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207696.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207696.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207696.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207696.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207696.hg.1 |
TC0100014377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100207697.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207697.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207697.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100207697.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100207697.hg.1 |
TC0100014377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000418662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000421455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
ENST00000438559.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014377.hg.1 |
NR_110618.1 |
Refseq |