Assay Details
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016120.3 | NP_057204.2 | 4 | 5858 | 156 | |
NM_183353.2 | NP_899196.1 | 5 | 5920 | 156 | ||
GenBank mRNA | AK123088.1 | - | 1 | 1654 | 156 | |
AK124877.1 | - | 1 | 1016 | 156 | ||
BX648613.1 | - | 1 | 3176 | 156 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X001162.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
random inactivation of X chromosome
regulation of dosage compensation by inactivation of X chromosome
transcription corepressor activity
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity
negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
random inactivation of X chromosome
regulation of dosage compensation by inactivation of X chromosome
transcription corepressor activity
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009538.hg.1 | ENST00000434255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009538.hg.1 | ENST00000434255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009397.hg.1 | ENST00000585082.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500011722.hg.1 | ENST00000579867.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | ENST00000469174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X015659.hg.1 | TC0X001162.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X015659.hg.1 | ENST00000332687.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X015659.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X015659.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00168884.hg.1 | TC0X00010097.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500011722.hg.1 | ENST00000579867.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010484.hg.1 | ENST00000561674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009397.hg.1 | ENST00000585082.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000332687.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000349225.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | ENST00000469174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_016120.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010097.hg.1 | NM_183353.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009538.hg.1 | ENST00000434255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009538.hg.1 | ENST00000434255.1 | Ensembl |
