Assay Details
| Gene Symbol: CASP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001224.4 | NP_001215.1 | 9 - 10 | 1029 | 64 | |
| GenBank mRNA | U13022.1 | - | 9 - 10 | 1029 | 64 | |
More Information
Set Membership: |
|

CASP2
Genomic Map