Assay Details
| Gene Symbol: CLCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000083.2 | NP_000074.2 | 14 - 15 | 1673 | 108 | |
| NR_046453.1 | 13 - 14 | 1526 | 108 | |||
| XM_011515782.2 | XP_011514084.1 | 4 - 5 | 421 | 108 | ||
| XM_017011739.1 | XP_016867228.1 | 11 - 12 | 1398 | 108 | ||
| XM_017011740.1 | XP_016867229.1 | 11 - 12 | 1374 | 108 | ||
| GenBank mRNA | BC112156.1 | - | 14 - 15 | 1638 | 8327624 | 108 |
| BC113495.1 | - | 14 - 15 | 1638 | 8322547 | 108 | |
| BC143796.1 | - | 13 - 14 | 1491 | 108 | ||
| M97820.1 | - | 11 - 12 | 1077 | 108 | ||
| Z25884.1 | - | 14 - 15 | 1673 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000934.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014444.hg.1 | TC07000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014444.hg.1 | ENST00000343257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014444.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167462.hg.1 | TC0700009491.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |

CLCN1
Genomic Map