Assay Details
| Gene Symbol: CLCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000083.2 | NP_000074.2 | 19 - 20 | 2448 | 76 | |
| NR_046453.1 | 18 - 19 | 2301 | 76 | |||
| XM_011515782.2 | XP_011514084.1 | 9 - 10 | 1196 | 76 | ||
| XM_017011739.1 | XP_016867228.1 | 16 - 17 | 2173 | 76 | ||
| XM_017011740.1 | XP_016867229.1 | 16 - 17 | 2149 | 76 | ||
| GenBank mRNA | BC112156.1 | - | 19 - 20 | 2413 | 8327624 | 76 |
| BC113495.1 | - | 19 - 20 | 2413 | 8322547 | 76 | |
| BC143796.1 | - | 18 - 19 | 2266 | 76 | ||
| M97820.1 | - | 16 - 17 | 1852 | 76 | ||
| Z25884.1 | - | 19 - 20 | 2448 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000934.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014449.hg.1 | TC07000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014449.hg.1 | ENST00000343257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014449.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167467.hg.1 | TC0700009491.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000343257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000432192.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000455478.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | ENST00000495612.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NM_000083.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009491.hg.1 | NR_046453.1 | Refseq |

CLCN1
Genomic Map