Assay Details
Gene Symbol: LMX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174069.1 | NP_001167540.1 | 2 - 3 | 273 | 64 | |
NM_177398.3 | NP_796372.1 | 2 - 3 | 462 | 64 | ||
XM_011509540.2 | XP_011507842.1 | 2 - 3 | 462 | 64 | ||
GenBank mRNA | AK122800.1 | - | 2 - 3 | 273 | 64 | |
AK127724.1 | - | 2 - 3 | 462 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003463.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
transcription, DNA-templated
axon guidance
dentate gyrus development
cerebellum development
negative regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
dopaminergic neuron differentiation
midbrain dopaminergic neuron differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
protein binding
zinc ion binding
regulation of cell growth
transcription, DNA-templated
axon guidance
dentate gyrus development
cerebellum development
negative regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
dopaminergic neuron differentiation
midbrain dopaminergic neuron differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053910.hg.1 | TC01003463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053910.hg.1 | ENST00000294816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053910.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | TC01003463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | ENST00000294816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | ENST00000342310.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053904.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | TC01003463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | ENST00000294816.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | ENST00000342310.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053905.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01053911.hg.1 | TC01003463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01053911.hg.1 | ENST00000342310.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01053911.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223681.hg.1 | TC0100016252.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100223686.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223686.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223686.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223686.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223686.hg.1 | TC0100016252.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100223687.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223687.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223687.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223687.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223687.hg.1 | TC0100016252.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | ENST00000489443.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100016252.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000294816.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000342310.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | ENST00000367893.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | NM_001174069.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | NM_177398.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100223682.hg.1 | TC0100016252.hg.1 | Clariom D TC |
