Assay Details
| Gene Symbol: EDARADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080738.3 | NP_542776.1 | 1 - 2 | 245 | 67 | |
| GenBank mRNA | AK096339.1 | - | 1 - 2 | 245 | 67 | |
| AK291930.1 | - | 1 - 2 | 160 | 67 | ||
| AK314634.1 | - | 1 - 2 | 417 | 67 | ||
| AY028914.1 | - | 1 - 2 | 229 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001953.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK291930.1,AK314634.1,AY028914.1,NM_080738.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01030132.hg.1 | TC01001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030132.hg.1 | ENST00000359362.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030132.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030133.hg.1 | TC01001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030133.hg.1 | ENST00000334232.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030133.hg.1 | ENST00000359362.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030133.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030133.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188984.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188984.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188984.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188984.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188984.hg.1 | TC0100012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188986.hg.1 | TC0100012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100188985.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188985.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188985.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188985.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188985.hg.1 | TC0100012093.hg.1 | Clariom D TC |

EDARADD
Genomic Map