Assay Details
| Gene Symbol: SOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005633.3 | NP_005624.2 | 12 - 13 | 2102 | 89 | |
| XM_005264515.4 | XP_005264572.1 | 12 - 13 | 2331 | 89 | ||
| XM_011533062.2 | XP_011531364.1 | 12 - 13 | 2270 | 89 | ||
| XM_011533064.2 | XP_011531366.1 | 12 - 13 | 2010 | 89 | ||
| XM_011533066.2 | XP_011531368.1 | 5 - 6 | 1011 | 89 | ||
| GenBank mRNA | AB209140.1 | - | 5 - 6 | 1073 | 89 | |
| AK290228.1 | - | 13 - 14 | 2148 | 89 | ||
| L13857.1 | - | 12 - 13 | 2061 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001761.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
B cell homeostasis
hair follicle development
cardiac atrium morphogenesis
pericardium morphogenesis
signal transduction
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
vitellogenesis
axon guidance
insulin receptor signaling pathway
regulation of T cell differentiation in thymus
regulation of Rho protein signal transduction
multicellular organism growth
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
regulation of T cell proliferation
positive regulation of apoptotic process
positive regulation of GTPase activity
positive regulation of epidermal growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
blood vessel morphogenesis
leukocyte migration
regulation of small GTPase mediated signal transduction
positive regulation of small GTPase mediated signal transduction
palate development
eyelid development in camera-type eye
heart trabecula morphogenesis
midbrain morphogenesis
regulation of pro-B cell differentiation
DNA binding
guanyl-nucleotide exchange factor activity
Ras guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
SH3 domain binding
protein heterodimerization activity
MAPK cascade
B cell homeostasis
hair follicle development
cardiac atrium morphogenesis
pericardium morphogenesis
signal transduction
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
vitellogenesis
axon guidance
insulin receptor signaling pathway
regulation of T cell differentiation in thymus
regulation of Rho protein signal transduction
multicellular organism growth
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
regulation of T cell proliferation
positive regulation of apoptotic process
positive regulation of GTPase activity
positive regulation of epidermal growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
blood vessel morphogenesis
leukocyte migration
regulation of small GTPase mediated signal transduction
positive regulation of small GTPase mediated signal transduction
palate development
eyelid development in camera-type eye
heart trabecula morphogenesis
midbrain morphogenesis
regulation of pro-B cell differentiation
DNA binding
guanyl-nucleotide exchange factor activity
Ras guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity
protein binding
SH3 domain binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000395038.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000402219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000426016.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000451331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000451331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000451331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000451331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000461545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000461545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000461545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000461545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000469581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000469581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000469581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000472480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000472480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000474390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000474390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000474390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | ENST00000474390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012307.hg.1 | NM_005633.3 | Refseq |

SOS1
Genomic Map