Assay Details
Gene Symbol: IZUMO1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199206.1 | NP_001186135.1 | 3 - 4 | 487 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000918.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P206702 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11011501.hg.1 | ENST00000440961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11011501.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11011501.hg.1 | TC11000918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11011502.hg.1 | ENST00000440961.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11011502.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11011502.hg.1 | TC11000918.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008800.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164998.hg.1 | TC1100008800.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100164999.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164999.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164999.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164999.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164999.hg.1 | TC1100008800.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | ENST00000328458.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | ENST00000440961.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | NM_001199206.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100164997.hg.1 | TC1100008800.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046872 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046872 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3243846 | NM_001199206.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3243846 | NM_001199206.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1665902 | NM_001199206.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2279217 | NM_001199206.1 | human_transcriptome |
