Assay Details
Gene Symbol: DHX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164239.1 | NP_001157711.1 | 2 - 3 | 334 | 81 | |
NM_003587.4 | NP_003578.2 | 2 - 3 | 655 | 81 | ||
XM_011514940.2 | XP_011513242.1 | 1 - 2 | 187 | 81 | ||
GenBank mRNA | AB001601.1 | - | 2 - 3 | 582 | 81 | |
AK302721.1 | - | 2 - 3 | 328 | 81 | ||
AK312264.1 | - | 2 - 3 | 594 | 81 | ||
BC008825.2 | - | 2 - 3 | 606 | 4100320 | 81 | |
BC009392.2 | - | 2 - 3 | 606 | 4136069 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001504.hg.1, TC6_apd_hap1000074.hg.1, TC6_cox_hap2000138.hg.1, TC6_dbb_hap3000127.hg.1, TC6_mann_hap4000116.hg.1, TC6_mcf_hap5000117.hg.1, TC6_qbl_hap6000128.hg.1, TC6_ssto_hap7000119.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011514940.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002406.hg.1 | TC6_qbl_hap6000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002406.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002406.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002433.hg.1 | TC6_cox_hap2000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002433.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002433.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002363.hg.1 | TC6_dbb_hap3000127.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002363.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002363.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002404.hg.1 | TC6_qbl_hap6000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002404.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002404.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002205.hg.1 | TC6_ssto_hap7000119.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002205.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002205.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001026.hg.1 | TC6_apd_hap1000074.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001026.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001026.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002428.hg.1 | TC6_cox_hap2000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002428.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002428.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002358.hg.1 | TC6_dbb_hap3000127.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002358.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002358.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002361.hg.1 | TC6_dbb_hap3000127.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002361.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002361.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002431.hg.1 | TC6_cox_hap2000138.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002431.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002431.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001951.hg.1 | TC6_mann_hap4000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001951.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001951.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002118.hg.1 | TC6_mcf_hap5000117.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002118.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002118.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001948.hg.1 | TC6_mann_hap4000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001948.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001948.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002202.hg.1 | TC6_ssto_hap7000119.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002202.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002202.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002207.hg.1 | TC6_ssto_hap7000119.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002207.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002207.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002401.hg.1 | TC6_qbl_hap6000128.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002401.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002401.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016990.hg.1 | TC06001504.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06016990.hg.1 | ENST00000376442.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06016990.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06016990.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001953.hg.1 | TC6_mann_hap4000116.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001953.hg.1 | NM_001164239.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001953.hg.1 | NM_003587.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376437.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600011375.hg.1 | ENST00000480966.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176682.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176682.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176682.hg.1 | ENST00000376442.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176682.hg.1 | ENST00000415603.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600176682.hg.1 | TC0600011375.hg.1 | Clariom D TC |
