Assay Details
| Gene Symbol: NSUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022072.3 | NP_071355.1 | 1 - 2 | 133 | 58 | |
| GenBank mRNA | AF169972.1 | - | 1 - 2 | 141 | 58 | |
| AK026262.1 | - | 1 - 2 | 125 | 58 | ||
| BC020602.1 | - | 1 - 2 | 133 | 4713693 | 58 | |
| BC057238.1 | - | 1 - 2 | 137 | 6463177 | 58 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000475.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF169972.1,AK026262.1,BC020602.1,BC057238.1,NM_022072.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03009140.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009140.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009140.hg.1 | ENST00000485793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009140.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009141.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009141.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009141.hg.1 | ENST00000485793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009141.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009145.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009145.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009145.hg.1 | ENST00000485793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009145.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157414.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157420.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157415.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |

NSUN3
Genomic Map