Assay Details
| Gene Symbol: NSUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022072.3 | NP_071355.1 | 2 - 3 | 237 | 67 | |
| GenBank mRNA | AF169972.1 | - | 2 - 3 | 245 | 67 | |
| AK026262.1 | - | 2 - 3 | 229 | 67 | ||
| BC020602.1 | - | 2 - 3 | 237 | 4713693 | 67 | |
| BC057238.1 | - | 2 - 3 | 241 | 6463177 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000475.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF169972.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03009150.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009150.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009150.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009151.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009151.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009151.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009146.hg.1 | TC03000475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009146.hg.1 | ENST00000314622.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009146.hg.1 | ENST00000485793.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009146.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157421.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157425.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300157426.hg.1 | TC0300008047.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000314622.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000461625.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000468555.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000476588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000477077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000483378.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000485793.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | ENST00000494128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008047.hg.1 | NM_022072.4 | Refseq |

NSUN3
Genomic Map