Assay Details
| Gene Symbol: GSTTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003081.1 | 1 - 2 | 117 | 148 | ||
| GenBank mRNA | AL050257.1 | - | 1 - 2 | 117 | 148 | |
| BC096726.1 | - | 1 - 2 | 117 | 148 | ||
| BC098364.1 | - | 1 - 2 | 117 | 148 | ||
| BC099710.1 | - | 1 - 2 | 117 | 148 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
|---|
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000581.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22010272.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010272.hg.1 | TC22000581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010274.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22010274.hg.1 | TC22000581.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | TC2200008188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158670.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | TC2200008188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158671.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008188.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | TC2200008188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | NR_003082.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000611600.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000612717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000612817.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000617461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000617532.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | ENST00000621179.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | NR_003081.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200158667.hg.1 | NR_003082.1 | Refseq |

GSTTP1
Genomic Map