Assay Details
| Gene Symbol: GALNT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253826.1 | NP_001240755.1 | 3 - 4 | 952 | 88 | |
| GenBank mRNA | AK022753.1 | - | 3 - 4 | 373 | 88 | |
More Information
Set Membership: |
|

GALNT14
Genomic Map