Assay Details
| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147128.3 | NP_667339.1 | 2 - 3 | 1618 | 124 | |
| GenBank mRNA | AF513707.1 | - | 2 - 3 | 1618 | 124 | |
| AF527533.1 | - | 2 - 3 | 945 | 124 | ||
| AY054122.1 | - | 3 - 4 | 382 | 124 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003390.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07003013.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003013.hg.1 | ENST00000319243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003013.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003013.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003015.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003015.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003015.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003019.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003019.hg.1 | ENST00000319243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003019.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003019.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003012.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003012.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003012.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003014.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003014.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003014.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003017.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003017.hg.1 | ENST00000319243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003017.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003017.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003017.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003020.hg.1 | TC07003390.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003020.hg.1 | ENST00000319243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003020.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07003020.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000426767.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000442865.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | ENST00000454624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_024278.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010614.hg.1 | NR_027347.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196481.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196481.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196481.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196481.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196481.hg.1 | TC0700013343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196485.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196485.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196485.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196485.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196485.hg.1 | TC0700013343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196479.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196479.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196479.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196479.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196479.hg.1 | TC0700013343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196482.hg.1 | TC0700013343.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | ENST00000459998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013343.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196484.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196484.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196484.hg.1 | ENST00000323037.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700196484.hg.1 | NM_147128.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700196484.hg.1 | TC0700013343.hg.1 | Clariom D TC |

ZNRF2
Genomic Map