Assay Details
| Gene Symbol: CRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173689.6 | NP_775960.4 | 4 - 5 | 846 | 137 | |
| XM_011518556.2 | XP_011516858.1 | 4 - 5 | 846 | 137 | ||
| XM_011518557.2 | XP_011516859.1 | 4 - 5 | 848 | 137 | ||
| XM_011518558.2 | XP_011516860.1 | 5 - 6 | 1208 | 137 | ||
| GenBank mRNA | AK123000.1 | - | 4 - 5 | 836 | 137 | |
| AY720432.1 | - | 4 - 5 | 752 | 137 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000629.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mesoderm formation
somitogenesis
positive regulation of epithelial to mesenchymal transition
negative regulation of endopeptidase activity
notochord formation
positive regulation of BMP signaling pathway
maintenance of epithelial cell apical/basal polarity
retina morphogenesis in camera-type eye
cardiovascular system development
calcium ion binding
aspartic-type endopeptidase inhibitor activity
enzyme binding
mesoderm formation
somitogenesis
positive regulation of epithelial to mesenchymal transition
negative regulation of endopeptidase activity
notochord formation
positive regulation of BMP signaling pathway
maintenance of epithelial cell apical/basal polarity
retina morphogenesis in camera-type eye
cardiovascular system development
calcium ion binding
aspartic-type endopeptidase inhibitor activity
enzyme binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09007249.hg.1 | TC09000629.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007249.hg.1 | ENST00000359999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007249.hg.1 | ENST00000373629.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007249.hg.1 | ENST00000373631.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007249.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007245.hg.1 | TC09000629.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007245.hg.1 | ENST00000359999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007245.hg.1 | ENST00000373631.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007245.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007246.hg.1 | TC09000629.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007246.hg.1 | ENST00000359999.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007246.hg.1 | ENST00000373631.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007246.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158052.hg.1 | TC0900008694.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158054.hg.1 | TC0900008694.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158051.hg.1 | TC0900008694.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000359999.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000373631.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | ENST00000460253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NM_173689.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008694.hg.1 | NR_104603.1 | Refseq |

CRB2
Genomic Map