Assay Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280787.1 | NP_001267716.1 | 5 - 6 | 568 | 119 | |
| NM_003172.3 | NP_003163.1 | 6 - 7 | 620 | 119 | ||
| XM_011518942.1 | XP_011517244.1 | 5 - 6 | 670 | 119 | ||
| GenBank mRNA | AK291122.1 | - | 6 - 7 | 603 | 119 | |
| BC028314.1 | - | 6 - 7 | 608 | 5190921 | 119 | |
| BC071658.1 | - | 6 - 7 | 625 | 5539506 | 119 | |
| KU178341.1 | - | 6 - 7 | 588 | 119 | ||
| KU178342.1 | - | 5 - 6 | 536 | 119 | ||
| KU178343.1 | - | 4 - 5 | 402 | 119 | ||
| Z35093.1 | - | 6 - 7 | 602 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001687.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09021619.hg.1 | TC09001687.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021619.hg.1 | ENST00000371974.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021619.hg.1 | ENST00000495952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021619.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021618.hg.1 | TC09001687.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021618.hg.1 | ENST00000371974.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021618.hg.1 | ENST00000495952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021618.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181050.hg.1 | TC0900011841.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000371974.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000437995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000463965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000495952.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000615505.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | ENST00000626556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_001280787.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011841.hg.1 | NM_003172.3 | Refseq |

SURF1
Genomic Map