Assay Details
| Gene Symbol: SLC5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003041.3 | NP_003032.1 | 9 - 10 | 1147 | 56 | |
| XM_006721072.3 | XP_006721135.2 | 9 - 10 | 1730 | 56 | ||
| GenBank mRNA | M95549.1 | - | 9 - 10 | 1148 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000379.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000330498.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000419665.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000562006.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000564197.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000565446.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000567051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000568891.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | ENST00000569576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NM_003041.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007538.hg.1 | NR_130783.1 | Refseq |

SLC5A2
Genomic Map