Assay Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130001.2 | NP_001123473.1 | 3 - 4 | 402 | 77 | |
GenBank mRNA | BC010062.2 | - | 3 - 4 | 302 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000863.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03015210.hg.1 | TC03000863.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03015210.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015210.hg.1 | ENST00000449199.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03015210.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03015210.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300166677.hg.1 | TC0300009302.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000312275.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000426338.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000449199.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000459838.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000461040.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000468043.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | ENST00000498159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001099777.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130001.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001130002.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NM_001243729.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009302.hg.1 | NR_024016.2 | Refseq |
