Assay Details
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018010.3 | NP_060480.1 | 1 - 2 | 459 | 57 | |
| GenBank mRNA | AB014762.1 | - | 1 - 2 | 211 | 57 | |
| AF139576.1 | - | 1 - 2 | 289 | 57 | ||
| AF245220.1 | - | 1 - 2 | 353 | 57 | ||
| AK001009.1 | - | 1 - 2 | 298 | 57 | ||
| BC011899.2 | - | 1 - 2 | 308 | 4053749 | 57 | |
| CR457251.1 | - | 1 - 2 | 211 | 57 | ||
| DA362330.1 | - | 1 - 2 | 459 | 57 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001630.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029426.hg.1 | TC03001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029426.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029426.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029420.hg.1 | TC03001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029420.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029420.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029419.hg.1 | TC03001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029419.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029419.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029422.hg.1 | TC03001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029422.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029422.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029424.hg.1 | TC03001630.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029424.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029424.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188842.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188843.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188845.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188840.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188839.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300188847.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188847.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188847.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300188847.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300188847.hg.1 | TC0300011939.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 171023 | - | - |

IFT57
Genomic Map