Assay Details
| Gene Symbol: IFT57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018010.3 | NP_060480.1 | 7 - 8 | 1091 | 103 | |
| GenBank mRNA | AB014762.1 | - | 7 - 8 | 843 | 103 | |
| AF139576.1 | - | 7 - 8 | 921 | 103 | ||
| AF245220.1 | - | 7 - 8 | 985 | 103 | ||
| AK001009.1 | - | 7 - 8 | 930 | 103 | ||
| AK298854.1 | - | 7 - 8 | 989 | 103 | ||
| BC009690.2 | - | 3 - 4 | 258 | 103 | ||
| BC011899.2 | - | 7 - 8 | 940 | 4053749 | 103 | |
| CR457251.1 | - | 7 - 8 | 843 | 103 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
neural tube closure
heart looping
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
smoothened signaling pathway
intraciliary transport
cilium assembly
regulation of apoptotic process
negative regulation of epithelial cell proliferation
left/right pattern formation
DNA binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007172.hg.1 | ENST00000457963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007172.hg.1 | ENST00000457963.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000264538.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000465024.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000468021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000478157.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000485979.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | ENST00000492106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011939.hg.1 | NM_018010.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007172.hg.1 | ENST00000457963.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007172.hg.1 | ENST00000457963.2 | Ensembl |

IFT57
Genomic Map