Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000003583.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000337248.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000374409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000468303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000475760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000483528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000498488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199013.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_178122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000003583.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000337248.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000374409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000468303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000475760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000483528.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
ENST00000498488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199012.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199013.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_001199014.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
NM_178122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198936.hg.1 |
TC0100013323.hg.1 |
Clariom D TC |