Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270894.1 | NP_001257823.1 | 3 - 4 | 401 | 94 | |
| NM_001270895.1 | NP_001257824.1 | 3 - 4 | 370 | 94 | ||
| NM_001270896.1 | NP_001257825.1 | 3 - 4 | 622 | 94 | ||
| NM_014408.4 | NP_055223.1 | 3 - 4 | 377 | 94 | ||
| GenBank mRNA | AF041432.1 | - | 3 - 4 | 332 | 94 | |
| AJ224335.1 | - | 3 - 4 | 277 | 94 | ||
| AK094755.1 | - | 2 - 3 | 1031 | 94 | ||
| AK315610.1 | - | 3 - 4 | 353 | 94 | ||
| AY007139.1 | - | 3 - 4 | 311 | 94 | ||
| BC007662.2 | - | 3 - 4 | 329 | 3051295 | 94 | |
| BG704657.1 | - | 3 - 4 | 382 | 94 | ||
| BI462429.1 | - | 3 - 4 | 604 | 94 | ||
| BI762992.1 | - | 3 - 4 | 372 | 94 | ||
| BM451581.1 | - | 3 - 4 | 370 | 94 | ||
| CD251784.1 | - | 3 - 4 | 419 | 94 | ||
More Information
Set Membership: |
|

TRAPPC3
Genomic Map