Assay Details
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK290288.1 | - | 2 - 3 | 249 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000217425.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000339946.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000342873.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000372676.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000447118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000462062.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | ENST00000488143.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007492.hg.1 | NM_006103.3 | Refseq |
