Assay Details
| Gene Symbol: PCTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102402.2 | NP_001095872.1 | 2 - 3 | 494 | 108 | |
| NM_021213.3 | NP_067036.2 | 2 - 3 | 387 | 108 | ||
| XM_005257549.1 | XP_005257606.1 | 2 - 3 | 387 | 108 | ||
| XM_005257550.3 | XP_005257607.1 | 2 - 3 | 387 | 108 | ||
| XM_017024913.1 | XP_016880402.1 | 2 - 3 | 469 | 108 | ||
| GenBank mRNA | AF087857.1 | - | 2 - 3 | 372 | 108 | |
| AF114430.1 | - | 2 - 3 | 277 | 108 | ||
| AF151638.1 | - | 2 - 3 | 371 | 108 | ||
| AK024667.1 | - | 2 - 3 | 108 | |||
| AK058120.1 | - | 2 - 3 | 320 | 108 | ||
| AK314509.1 | - | 2 - 3 | 313 | 108 | ||
| BC005112.1 | - | 2 - 3 | 199 | 108 | ||
| BC012084.1 | - | 2 - 3 | 305 | 4575824 | 108 | |
| BM153142.1 | - | 2 - 3 | 387 | 108 | ||
| BM549250.1 | - | 2 - 3 | 494 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000695.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001102402.2,XM_017024913.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | ENST00000268896.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | ENST00000325214.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | ENST00000417982.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17009116.hg.1 | TC17000695.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162178.hg.1 | TC1700008342.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700162176.hg.1 | TC1700008342.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |

PCTP
Genomic Map