Assay Details
| Gene Symbol: PCTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005257549.1 | XP_005257606.1 | 5 - 6 | 707 | 78 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000268896.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000325214.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000417982.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000571489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000572536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000573500.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576183.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | ENST00000576221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_001102402.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008342.hg.1 | NM_021213.3 | Refseq |

PCTP
Genomic Map