Assay Details
| Gene Symbol: PDIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006810.3 | NP_006801.1 | 11 - 12 | 1063 | 66 | |
| GenBank mRNA | D49490.1 | - | 11 - 12 | 963 | 66 | |
More Information
Set Membership: |
|

PDIA5
Genomic Map