Assay Details
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152434.2 | NP_689647.2 | 14 - 15 | 2229 | 140 | |
XM_017017239.1 | XP_016872728.1 | 13 - 14 | 1547 | 140 | ||
GenBank mRNA | AK056905.1 | - | 12 - 13 | 1881 | 140 | |
AK126881.1 | - | 14 - 15 | 2220 | 140 | ||
AK293001.1 | - | 14 - 15 | 2164 | 140 | ||
BC056241.1 | - | 2 - 3 | 324 | 140 | ||
BC110440.1 | - | 11 - 12 | 1842 | 40001348 | 139 | |
BC110441.1 | - | 11 - 12 | 1784 | 40001349 | 140 | |
BC110442.1 | - | 11 - 12 | 1842 | 40001350 | 140 | |
BC118669.1 | - | 11 - 12 | 1845 | 40012545 | 140 | |
BX647832.1 | - | 14 - 15 | 2203 | 140 | ||
BX648704.1 | - | 14 - 15 | 2208 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002259.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11027011.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027011.hg.1 | ENST00000409771.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027011.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027011.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027013.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027013.hg.1 | ENST00000409771.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027013.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027013.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193857.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
