Assay Details
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152434.2 | NP_689647.2 | 17 - 18 | 2569 | 95 | |
XM_017017239.1 | XP_016872728.1 | 16 - 17 | 1887 | 95 | ||
GenBank mRNA | AK056905.1 | - | 15 - 16 | 2221 | 95 | |
AK126881.1 | - | 17 - 18 | 2560 | 95 | ||
AK293001.1 | - | 17 - 18 | 2504 | 95 | ||
BC056241.1 | - | 5 - 6 | 664 | 95 | ||
BC110439.1 | - | 15 - 16 | 2259 | 40001346 | 95 | |
BC110440.1 | - | 14 - 15 | 2181 | 40001348 | 95 | |
BC110441.1 | - | 14 - 15 | 2124 | 40001349 | 95 | |
BC110442.1 | - | 14 - 15 | 2182 | 40001350 | 95 | |
BC118669.1 | - | 14 - 15 | 2185 | 40012545 | 95 | |
BX647832.1 | - | 17 - 18 | 2543 | 95 | ||
BX648704.1 | - | 17 - 18 | 2548 | 95 | ||
BX649022.1 | - | 17 - 18 | 2539 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002259.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027008.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027008.hg.1 | ENST00000409771.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027008.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027008.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193854.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193853.hg.1 | TC1100012208.hg.1 | Clariom D TC |
