Assay Details
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152434.2 | NP_689647.2 | 1 - 2 | 131 | 91 | |
XM_011542620.2 | XP_011540922.1 | 1 - 2 | 129 | 91 | ||
XM_017017239.1 | XP_016872728.1 | 1 - 2 | 102 | 91 | ||
GenBank mRNA | AK126881.1 | - | 1 - 2 | 124 | 91 | |
AK293001.1 | - | 1 - 2 | 65 | 91 | ||
BC070142.1 | - | 1 - 2 | 110 | 91 | ||
BC107418.1 | - | 1 - 2 | 113 | 91 | ||
BX647832.1 | - | 1 - 2 | 105 | 91 | ||
BX648704.1 | - | 1 - 2 | 110 | 91 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002259.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293001.1,XM_017017239.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11027034.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027034.hg.1 | ENST00000433523.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027034.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027034.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11027035.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027035.hg.1 | ENST00000433523.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027035.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027035.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193883.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193883.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193883.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193883.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193883.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100193882.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193882.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193882.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193882.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193882.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100193884.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193884.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193884.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193884.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193884.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 105348 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 116902 | - | - |
