Assay Details
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152434.2 | NP_689647.2 | 2 - 3 | 243 | 185 | |
XM_011542620.2 | XP_011540922.1 | 2 - 3 | 241 | 185 | ||
XM_017017239.1 | XP_016872728.1 | 2 - 3 | 214 | 185 | ||
GenBank mRNA | AK126881.1 | - | 2 - 3 | 236 | 185 | |
AK293001.1 | - | 2 - 3 | 177 | 186 | ||
BC070142.1 | - | 2 - 3 | 222 | 185 | ||
BC107418.1 | - | 2 - 3 | 225 | 186 | ||
BX647832.1 | - | 2 - 3 | 217 | 185 | ||
BX648704.1 | - | 2 - 3 | 222 | 185 | ||
BX649022.1 | - | 2 - 3 | 213 | 185 | ||
DA291472.1 | - | 2 - 3 | 245 | 185 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002259.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293001.1,XM_017017239.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027033.hg.1 | ENST00000282251.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027033.hg.1 | ENST00000433523.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027033.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027033.hg.1 | TC11002259.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100193881.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193881.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193881.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100193881.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100193881.hg.1 | TC1100012208.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000282251.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000417747.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000431778.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000462890.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | ENST00000532251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012208.hg.1 | NM_152434.2 | Refseq |
