Assay Details
| Gene Symbol: EDARADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145861.2 | NP_665860.2 | 1 - 2 | 123 | 87 | |
| GenBank mRNA | AK290862.1 | - | 1 - 2 | 159 | 87 | |
| AY028913.1 | - | 1 - 2 | 123 | 87 | ||
| BC114557.1 | - | 1 - 2 | 78 | 87 | ||
| BC114560.1 | - | 1 - 2 | 78 | 87 | ||
| BC128082.1 | - | 1 - 2 | 78 | 40006675 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001953.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01030131.hg.1 | TC01001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030131.hg.1 | ENST00000334232.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030131.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030130.hg.1 | TC01001953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030130.hg.1 | ENST00000334232.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030130.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188983.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188983.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188983.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188983.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188983.hg.1 | TC0100012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100188981.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188981.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188981.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100188981.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100188981.hg.1 | TC0100012093.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000334232.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000359362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000366587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | ENST00000439430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_080738.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012093.hg.1 | NM_145861.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 145025 | - | - |

EDARADD
Genomic Map