Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
TC02004927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000361874.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000393999.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000394003.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000407639.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000409240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000409438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000409567.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
ENST00000409868.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_001135040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_001135041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_001190836.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_001190837.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_004082.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NM_023019.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02031818.hg.1 |
NR_033935.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000361874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000394003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409567.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000434055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000466110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000491465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000628224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000633691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001135040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001135041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001190836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001190837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_004082.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_023019.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NR_033935.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000361874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000394003.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409438.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409567.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000409868.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000434055.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000466110.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000491465.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000628224.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
ENST00000633691.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001135040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001135041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001190836.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_001190837.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_004082.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NM_023019.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
NR_033935.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200226886.hg.1 |
TC0200016690.hg.1 |
Clariom D TC |