Assay Details
| Gene Symbol: DET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135739.1 | 6 - 7 | 2287 | 64 | ||
| NR_135742.1 | 5 - 6 | 2426 | 64 | |||
| GenBank mRNA | BC073867.1 | - | 6 - 7 | 2229 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000268148.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000444300.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000557842.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000558413.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | ENST00000564406.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_001144074.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NM_017996.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010923.hg.1 | NR_026645.2 | Refseq |

DET1
Genomic Map