Assay Details
| Gene Symbol: AOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011944.1 | XP_016867433.1 | 2 - 3 | 214 | 72 | |
| XM_017011945.1 | XP_016867434.1 | 2 - 3 | 244 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017011944.1, XM_017011945.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
xenobiotic metabolic process
amine metabolic process
cellular response to copper ion starvation
response to drug
response to antibiotic
oxidation-reduction process
cellular response to copper ion
cellular response to histamine
cellular response to heparin
cellular response to azide
receptor activity
copper ion binding
calcium ion binding
primary amine oxidase activity
drug binding
heparin binding
zinc ion binding
protein complex binding
protein homodimerization activity
quinone binding
diamine oxidase activity
histamine oxidase activity
methylputrescine oxidase activity
propane-1,3-diamine oxidase activity
xenobiotic metabolic process
amine metabolic process
cellular response to copper ion starvation
response to drug
response to antibiotic
oxidation-reduction process
cellular response to copper ion
cellular response to histamine
cellular response to heparin
cellular response to azide
receptor activity
copper ion binding
calcium ion binding
primary amine oxidase activity
drug binding
heparin binding
zinc ion binding
protein complex binding
protein homodimerization activity
quinone binding
diamine oxidase activity
histamine oxidase activity
methylputrescine oxidase activity
propane-1,3-diamine oxidase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000360937.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000416793.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000460213.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000467291.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000480582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000483043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000493429.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000619575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | ENST00000622116.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001091.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009682.hg.1 | NM_001272072.1 | Refseq |

AOC1
Genomic Map