Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001185.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001185.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19001185.hg.1 |
TC19000073.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146513.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146513.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900146513.hg.1 |
TC1900006646.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900146513.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900146513.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
ENST00000322315.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006646.hg.1 |
NM_133261.2 |
Refseq |