Assay Details
Gene Symbol: RANBP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161429.2 | NP_001154901.1 | 8 - 9 | 1157 | 159 | |
NM_001323273.1 | NP_001310202.1 | 7 - 8 | 1075 | 159 | ||
NM_001323274.1 | NP_001310203.1 | 8 - 9 | 1150 | 159 | ||
NM_001323275.1 | NP_001310204.1 | 8 - 9 | 1090 | 159 | ||
NM_001323276.1 | NP_001310205.1 | 8 - 9 | 1090 | 159 | ||
NM_001323277.1 | NP_001310206.1 | 7 - 8 | 1075 | 159 | ||
NM_001323278.1 | NP_001310207.1 | 7 - 8 | 1015 | 159 | ||
NM_001323279.1 | NP_001310208.1 | 6 - 7 | 1003 | 159 | ||
NM_001323280.1 | NP_001310209.1 | 7 - 8 | 1015 | 159 | ||
NM_145000.4 | NP_659437.3 | 7 - 8 | 1082 | 159 | ||
XM_006714452.3 | XP_006714515.1 | 8 - 9 | 1184 | 159 | ||
XM_006714453.3 | XP_006714516.1 | 8 - 9 | 1124 | 159 | ||
XM_006714455.3 | XP_006714518.1 | 7 - 8 | 1049 | 159 | ||
GenBank mRNA | AK058151.1 | - | 7 - 8 | 1073 | 159 | |
BC047660.1 | - | 7 - 8 | 1075 | 5261736 | 159 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001271.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | TC05001271.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | ENST00000296604.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | ENST00000502994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | ENST00000515759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05017809.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000505865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010498.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000296604.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000502994.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | ENST00000515759.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | NM_001161429.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | NM_145000.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173116.hg.1 | TC0500010498.hg.1 | Clariom D TC |
