Assay Details
| Gene Symbol: NR1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021724.4 | NP_068370.1 | 5 - 6 | 1876 | 63 | |
| GenBank mRNA | AK300155.1 | - | 2 - 3 | 572 | 63 | |
| AK315270.1 | - | 6 - 7 | 1618 | 63 | ||
| BC047875.1 | - | 5 - 6 | 1700 | 6190140 | 63 | |
| BC056148.1 | - | 5 - 6 | 1767 | 6195844 | 63 | |
| HQ692861.1 | - | 5 - 6 | 1245 | 63 | ||
| M24898.1 | - | 5 - 6 | 1872 | 63 | ||
| X72631.1 | - | 5 - 6 | 1447 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK300155.1 transcripts.
Set Membership: |
|

NR1D1
Genomic Map