Assay Details
| Gene Symbol: NIPSNAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202502.1 | NP_001189431.1 | 2 - 3 | 183 | 75 | |
| NM_003634.3 | NP_003625.2 | 2 - 3 | 484 | 75 | ||
| GenBank mRNA | AJ001258.1 | - | 2 - 3 | 483 | 75 | |
| AK098367.1 | - | 2 - 3 | 238 | 75 | ||
| AK298816.1 | - | 2 - 3 | 185 | 75 | ||
| AK314407.1 | - | 2 - 3 | 255 | 75 | ||
| BC002371.2 | - | 2 - 3 | 234 | 2822369 | 75 | |
| BC006473.1 | - | 2 - 3 | 234 | 2822369 | 75 | |
| BC018869.2 | - | 1 - 2 | 126 | 75 | ||
| DA133702.1 | - | 2 - 3 | 302 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22001488.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22011151.hg.1 | TC22001488.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011151.hg.1 | ENST00000216121.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011151.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011151.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011152.hg.1 | TC22001488.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011152.hg.1 | ENST00000216121.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011152.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011152.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | TC2200009336.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | NM_003634.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000216121.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000415100.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000437094.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000455496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | ENST00000496944.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | NM_001202502.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200170163.hg.1 | NM_003634.3 | Refseq |

NIPSNAP1
Genomic Map