Assay Details
Gene Symbol: NIPSNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202502.1 | NP_001189431.1 | 4 - 5 | 324 | 70 | |
NM_003634.3 | NP_003625.2 | 4 - 5 | 625 | 70 | ||
GenBank mRNA | AJ001258.1 | - | 4 - 5 | 624 | 70 | |
AK098367.1 | - | 4 - 5 | 379 | 70 | ||
AK290953.1 | - | 4 - 5 | 378 | 70 | ||
AK298816.1 | - | 4 - 5 | 326 | 70 | ||
AK314407.1 | - | 4 - 5 | 396 | 70 | ||
BC002371.2 | - | 4 - 5 | 375 | 2822369 | 70 | |
BC006473.1 | - | 4 - 5 | 375 | 2822369 | 70 | |
BC018869.2 | - | 3 - 4 | 267 | 70 | ||
DA133702.1 | - | 4 - 5 | 443 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22001488.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011147.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011147.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011147.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011147.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011145.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011145.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011145.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011145.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170157.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170159.hg.1 | NM_003634.3 | Refseq |
