Assay Details
Gene Symbol: NIPSNAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003634.3 | NP_003625.2 | 1 - 2 | 347 | 115 | |
GenBank mRNA | AJ001258.1 | - | 1 - 2 | 346 | 115 | |
AK098367.1 | - | 1 - 2 | 101 | 115 | ||
AK290953.1 | - | 1 - 2 | 100 | 115 | ||
AK314407.1 | - | 1 - 2 | 118 | 115 | ||
BC002371.2 | - | 1 - 2 | 97 | 2822369 | 115 | |
BC006473.1 | - | 1 - 2 | 97 | 2822369 | 115 | |
DA133702.1 | - | 1 - 2 | 165 | 115 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22001488.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22011156.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011156.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011156.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011156.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011158.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011158.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011158.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011158.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700006690.hg.1 | ENST00000576249.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011165.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011165.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011165.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011153.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011153.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011153.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011153.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22011164.hg.1 | TC22001488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22011164.hg.1 | ENST00000216121.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22011164.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009336.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170169.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170175.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170175.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170175.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170175.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170175.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | NM_001202502.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170164.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000415100.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000455496.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000494966.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | ENST00000496944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170167.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700006690.hg.1 | ENST00000576249.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | TC2200009336.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | NM_003634.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | ENST00000216121.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | ENST00000437094.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200170176.hg.1 | NM_003634.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 202549 | - | - |
