Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
TC20001761.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000317619.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000317677.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000352393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000397442.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000397443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000397445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
ENST00000397446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_001198863.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_003915.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_152925.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_152926.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_152927.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NM_152928.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010872.hg.1 |
NR_037188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000317677.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000352393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000401607.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000430570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000437340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000483359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000483495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_001198863.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_003915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NR_037188.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
TC2000010007.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000317677.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000352393.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397443.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000397445.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000401607.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000430570.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000437340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000483359.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
ENST00000483495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_001198863.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_003915.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152925.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152926.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152927.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NM_152928.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171517.hg.1 |
NR_037188.1 |
Refseq |