Assay Details
Gene Symbol: MYLK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033118.3 | NP_149109.1 | 2 - 3 | 158 | 122 | |
GenBank mRNA | AF325549.1 | - | 1 - 2 | 56 | 122 | |
AJ272502.1 | - | 2 - 3 | 164 | 122 | ||
BC069627.1 | - | 1 - 2 | 56 | 7262446 | 122 | |
BC092413.1 | - | 2 - 3 | 158 | 30530456 | 122 | |
BC127622.1 | - | 1 - 2 | 56 | 40085085 | 122 | |
BX647635.1 | - | 2 - 3 | 141 | 122 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000199.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
striated muscle contraction
neuromuscular synaptic transmission
positive regulation of gene expression
skeletal muscle satellite cell differentiation
peptidyl-threonine phosphorylation
regulation of muscle filament sliding
skeletal muscle cell differentiation
protein autophosphorylation
cardiac muscle tissue morphogenesis
cardiac muscle contraction
calmodulin-dependent protein kinase activity
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
striated muscle contraction
neuromuscular synaptic transmission
positive regulation of gene expression
skeletal muscle satellite cell differentiation
peptidyl-threonine phosphorylation
regulation of muscle filament sliding
skeletal muscle cell differentiation
protein autophosphorylation
cardiac muscle tissue morphogenesis
cardiac muscle contraction
calmodulin-dependent protein kinase activity
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20002391.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002391.hg.1 | ENST00000375994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002391.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002391.hg.1 | TC20000199.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002390.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002390.hg.1 | ENST00000375994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002390.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002390.hg.1 | TC20000199.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002387.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002387.hg.1 | ENST00000375994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002387.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002387.hg.1 | TC20000199.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20002389.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002389.hg.1 | ENST00000375994.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20002389.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20002389.hg.1 | TC20000199.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | TC2000007091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148408.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | ENST00000468730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007091.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | TC2000007091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148406.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | TC2000007091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148407.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | NM_033118.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | TC2000007091.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | ENST00000375985.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | ENST00000375994.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000148404.hg.1 | NM_033118.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 183932 | - | - |
