Assay Details
| Gene Symbol: PLCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014996.2 | NP_055811.1 | 20 - 21 | 2607 | 126 | |
| XM_005247239.1 | XP_005247296.1 | 20 - 21 | 2964 | 126 | ||
| XM_011512564.2 | XP_011510866.1 | 21 - 22 | 2913 | 126 | ||
| XM_017005924.1 | XP_016861413.1 | 21 - 22 | 2913 | 126 | ||
| GenBank mRNA | AK022610.1 | - | 12 - 13 | 1416 | 126 | |
| AK096620.1 | - | 20 - 21 | 2964 | 126 | ||
| BC113950.1 | - | 20 - 21 | 2607 | 5501480 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PLCH1
Genomic Map